Learn More
Medchemexpress LLC Biotin-PEG4-hydrazide | 756525-97-0 | 99.9% | C21H39N5O7S | 25 MG
SDP
Catalog No. 5000352526
Encompass_Preferred
Biotin-PEG4-hydrazide is a biotin-labeled, PEG-based PROTAC linker. It is used in the synthesis of Proteolysis-targeting chimeras (PROTACs), which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52526 Supplier Medchemexpress LLC Supplier No. 25MGHY140932
Biotin-PEG4-hydrazide is a biotin-labeled, PEG-based PROTAC linker. It is used in the synthesis of Proteolysis-targeting chimeras (PROTACs), which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins