Learn More
Medchemexpress LLC Biotin-PEG4-hydrazide | 756525-97-0 | 99.9% | C21H39N5O7S | 50 MG
SDP
Catalog No. 5000355620
Encompass_Preferred
Biotin-PEG4-hydrazide is a biotin-labeled, PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are capable of exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins, making this product suitable for research applications.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-55620 Supplier Medchemexpress LLC Supplier No. HY14093250MG
Biotin-PEG4-hydrazide is a biotin-labeled, PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are capable of exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins, making this product suitable for research applications.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only