Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG4-methyltetrazine | 1962919-31-8 | 95.3% | 674.81 g·mol⁻¹ | C31H46N8O7S | 10 MG
SDP

Catalog No. 5000286755
Encompass_Preferred

Biotin-PEG4-MeTz is a biotinylated click-chemistry reagent containing a terminal methyltetrazine that reacts rapidly with trans-cyclooctene via inverse electron-demand Diels-Alder (iEDDA) chemistry. Supplied as a solid, it is intended for preparation of biotinylated conjugates and labeling applications in chemical biology and bioconjugation workflows.

  • Contains a terminal methyltetrazine for rapid iEDDA reactions.
  • Includes a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Facilitates preparation of biotinylated conjugates and biomolecule labeling.
  • Purple to purplish red solid; high purity (~95.3%).
  • Soluble in DMSO at about 250 mg/mL; ultrasonication recommended.
  • Store sealed at -20 °C away from moisture and light; in solution store at -80 °C for long term.

Catalog No. 50-002-86755 Supplier Medchemexpress LLC Supplier No. HY15174110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG4-MeTz is a biotinylated click-chemistry reagent containing a terminal methyltetrazine that reacts rapidly with trans-cyclooctene via inverse electron-demand Diels-Alder (iEDDA) chemistry. Supplied as a solid, it is intended for preparation of biotinylated conjugates and labeling applications in chemical biology and bioconjugation workflows.

  • Contains a terminal methyltetrazine for rapid iEDDA reactions.
  • Includes a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Facilitates preparation of biotinylated conjugates and biomolecule labeling.
  • Purple to purplish red solid; high purity (~95.3%).
  • Soluble in DMSO at about 250 mg/mL; ultrasonication recommended.
  • Store sealed at -20 °C away from moisture and light; in solution store at -80 °C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.