Learn More
Cayman Chemical Biotin-PEG4-NHS ester, Purity: ≥90%, CAS: 459426-22-3, 100 mg
SDP
Catalog No. 502941906
Encompass_Preferred
An amine reactive reagent; has been used in the biotinylation of proteins and antibodies
Catalog No. 50-294-1906 Supplier Cayman Chemical Supplier No. 29114100 MG
An amine reactive reagent; has been used in the biotinylation of proteins and antibodies