Learn More
Cayman Chemical BIotIn-PEG4-NHS 25mg
SDP
Catalog No. 502941910
Encompass_Preferred
An amine reactive reagent; has been used in the biotinylation of proteins and antibodies
Catalog No. 50-294-1910 Supplier Cayman Chemical Supplier No. 2911425 MG
An amine reactive reagent; has been used in the biotinylation of proteins and antibodies