Learn More
Medchemexpress LLC Biotin-PEG4-NHS ester | 459426-22-3 | 99.6% | C25H40N4O10S | 100 MG
SDP
Catalog No. 5000356300
Encompass_Preferred
Biotin-PEG4-NHS ester is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, enabling selective degradation of target proteins.
  • Biotin-labeled, PEG-based linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56300 Supplier Medchemexpress LLC Supplier No. HY140889100MG
Biotin-PEG4-NHS ester is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, enabling selective degradation of target proteins.
  • Biotin-labeled, PEG-based linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins