Learn More
Medchemexpress LLC Biotin-PEG4-NHS ester | 459426-22-3 | 99.6% | C25H40N4O10S | 5 G
SDP
Catalog No. 5000355160
Encompass_Preferred
Biotin-PEG4-NHS ester is a biotin-labeled, PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are compounds with two distinct ligands joined by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • High purity of 99.6%
  • Solid appearance, white to off-white color
  • Stable under recommended storage conditions
Catalog No. 50-003-55160 Supplier Medchemexpress LLC Supplier No. HY1408895G
Biotin-PEG4-NHS ester is a biotin-labeled, PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are compounds with two distinct ligands joined by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • High purity of 99.6%
  • Solid appearance, white to off-white color
  • Stable under recommended storage conditions