Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG4-OH | 1217609-84-1 | 98.0% | 100 MG
SDP

Catalog No. 5000339138
Encompass_Preferred

Biotin-PEG4-OH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It is intended for research use only and is not sold to patients.

  • Molecular weight of 419.54
  • Molecular formula of C18H33N3O6S
  • Appears as a solid
  • Color is white to off-white
  • Purity is 98.0%
  • Identified for laboratory chemicals and manufacturing
  • Stable under recommended storage conditions

Catalog No. 50-003-39138 Supplier Medchemexpress LLC Supplier No. HY130290100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG4-OH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It is intended for research use only and is not sold to patients.

  • Molecular weight of 419.54
  • Molecular formula of C18H33N3O6S
  • Appears as a solid
  • Color is white to off-white
  • Purity is 98.0%
  • Identified for laboratory chemicals and manufacturing
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.