Learn More
Medchemexpress LLC Biotin-PEG4-PFP ester | 1334172-58-5 | MFCD13184979 | 96.2% | 657.65 | C27H36F5N3O8S | 50 MG
SDP
Catalog No. 5000202693
Encompass_Preferred
Biotin-PEG4-PFP ester is an activated biotin linker containing a four-unit polyethylene glycol (PEG4) spacer and a pentafluorophenyl (PFP) ester leaving group. It is designed for efficient amine coupling and biotin tagging in bioconjugation workflows such as ADC and PROTAC synthesis.
  • Cleavable PEG4 spacer provides flexibility and solubility
  • PFP activated ester enables rapid coupling to primary amines
  • Biotin tag allows affinity capture and detection
  • Suitable for antibody-drug conjugate and probe labeling workflows
  • Stable as a powder at -20°C for extended storage
Catalog No. 50-002-02693 Supplier Medchemexpress LLC Supplier No. HY13848850MG
Biotin-PEG4-PFP ester is an activated biotin linker containing a four-unit polyethylene glycol (PEG4) spacer and a pentafluorophenyl (PFP) ester leaving group. It is designed for efficient amine coupling and biotin tagging in bioconjugation workflows such as ADC and PROTAC synthesis.
  • Cleavable PEG4 spacer provides flexibility and solubility
  • PFP activated ester enables rapid coupling to primary amines
  • Biotin tag allows affinity capture and detection
  • Suitable for antibody-drug conjugate and probe labeling workflows
  • Stable as a powder at -20°C for extended storage