Learn More
Medchemexpress LLC Biotin-PEG4-picolyl azide | 2222687-71-8 | MFCD29052186 | 98.7% | 622.74 | C27H42N8O7S | 5 MG
SDP
Catalog No. 5000223185
Encompass_Preferred
Biotin-PEG4-picolyl azide is a biotinylated click-chemistry reagent containing a PEG4 spacer and a picolyl azide moiety. It is designed for copper-catalyzed azide-alkyne cycloaddition (CuAAC) to conjugate biotin to alkyne-containing biomolecules for labeling, enrichment, and detection in chemical biology workflows.
  • Picolyl azide enhances copper-catalyzed azide-alkyne cycloaddition kinetics.
  • PEG4 spacer improves solubility and provides a flexible linkage.
  • Biotin tag enables affinity capture and detection of conjugates.
  • High purity suitable for analytical and preparative labeling workflows.
  • Small pack size appropriate for milligram-scale reactions.
Catalog No. 50-002-23185 Supplier Medchemexpress LLC Supplier No. HY1409155MG
Biotin-PEG4-picolyl azide is a biotinylated click-chemistry reagent containing a PEG4 spacer and a picolyl azide moiety. It is designed for copper-catalyzed azide-alkyne cycloaddition (CuAAC) to conjugate biotin to alkyne-containing biomolecules for labeling, enrichment, and detection in chemical biology workflows.
  • Picolyl azide enhances copper-catalyzed azide-alkyne cycloaddition kinetics.
  • PEG4 spacer improves solubility and provides a flexible linkage.
  • Biotin tag enables affinity capture and detection of conjugates.
  • High purity suitable for analytical and preparative labeling workflows.
  • Small pack size appropriate for milligram-scale reactions.