Learn More
Medchemexpress LLC Biotin-PEG4-TFP ester | 2985664-57-9 | 97.8% | C27H37F4N3O8S | 100 MG
SDP
Catalog No. 5000354566
Encompass_Preferred
Biotin-PEG4-TFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-54566 Supplier Medchemexpress LLC Supplier No. HY140903100MG
Biotin-PEG4-TFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins