Learn More
Medchemexpress LLC Biotin-PEG5-amine | 113072-75-6 | 96.24% | C22H42N4O7S | 100 MG
SDP
Catalog No. 5000354996
Encompass_Preferred
Biotin-PEG5-amine is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled, PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54996 Supplier Medchemexpress LLC Supplier No. HY140896100MG
Biotin-PEG5-amine is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled, PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins