Learn More
Medchemexpress LLC Biotin-PEG5-amine | 113072-75-6 | MFCD22574819 | >95.0% | C22H42N4O7S | 50 MG
SDP
Catalog No. 5000353385
Encompass_Preferred
Biotin-PEG5-amine is a biotinylation reagent reactive with NHS active ester for conjugation. It is a biotin-labeled, PEG-based PROTAC linker useful in the synthesis of PROTACs.
  • Reactive with NHS active ester for conjugation.
  • Biotin-labeled and PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
Catalog No. 50-003-53385 Supplier Medchemexpress LLC Supplier No. HY14089650MG
Biotin-PEG5-amine is a biotinylation reagent reactive with NHS active ester for conjugation. It is a biotin-labeled, PEG-based PROTAC linker useful in the synthesis of PROTACs.
  • Reactive with NHS active ester for conjugation.
  • Biotin-labeled and PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.