Learn More
Medchemexpress LLC Biotin-PEG6-alcohol | 906099-89-6 | 99.3% | C22H41N3O8S | 100 MG
SDP
Catalog No. 5000354878
Encompass_Preferred
Biotin-PEG6-alcohol is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that feature two distinct ligands connected by a linker. One ligand targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.
  • Biotin-labeled and PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Intended for research use only
Catalog No. 50-003-54878 Supplier Medchemexpress LLC Supplier No. HY140893100MG
Biotin-PEG6-alcohol is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that feature two distinct ligands connected by a linker. One ligand targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.
  • Biotin-labeled and PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Intended for research use only