Learn More
Medchemexpress LLC Biotin-PEG6-alcohol | 906099-89-6 | 99.3% | C22H41N3O8S | 50 MG
SDP
Catalog No. 5000355405
Encompass_Preferred
Biotin-PEG6-alcohol is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Biotin-labeled: This feature allows for specific targeting or detection in various biological assays due to biotin's high affinity for streptavidin.
  • PEG-based PROTAC linker: The polyethylene glycol (PEG) chain provides flexibility and enhances solubility, which are advantageous properties for the development of PROTACs (Proteolysis Targeting Chimeras).
  • Application in PROTAC synthesis: It functions as a key component in the creation of PROTACs.
  • Mechanism of PROTACs: PROTACs operate by harnessing the intracellular ubiquitin-proteasome system to achieve the selective degradation of target proteins.
Catalog No. 50-003-55405 Supplier Medchemexpress LLC Supplier No. HY14089350MG
Biotin-PEG6-alcohol is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Biotin-labeled: This feature allows for specific targeting or detection in various biological assays due to biotin's high affinity for streptavidin.
  • PEG-based PROTAC linker: The polyethylene glycol (PEG) chain provides flexibility and enhances solubility, which are advantageous properties for the development of PROTACs (Proteolysis Targeting Chimeras).
  • Application in PROTAC synthesis: It functions as a key component in the creation of PROTACs.
  • Mechanism of PROTACs: PROTACs operate by harnessing the intracellular ubiquitin-proteasome system to achieve the selective degradation of target proteins.