Learn More
Medchemexpress LLC Biotin-PEG6-azide | 1085938-09-5 | 99.9% | C24H44N6O8S | 100 MG
SDP
Catalog No. 5000353138
Encompass_Preferred
Biotin-PEG6-azide is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Designed for selective protein degradation via PROTACs
Catalog No. 50-003-53138 Supplier Medchemexpress LLC Supplier No. HY140911100MG
Biotin-PEG6-azide is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Designed for selective protein degradation via PROTACs