Learn More
Medchemexpress LLC Biotin-PEG6-azide | 1085938-09-5 | 99.9% | C24H44N6O8S | 25 MG
SDP
Catalog No. 5000357122
Encompass_Preferred
Biotin-PEG6-azide is a biotin-labeled, PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and functions as a click chemistry reagent, facilitating targeted protein degradation applications.
  • Biotin-labeled for detection and purification
  • PEG-based for improved solubility and reduced immunogenicity
  • PROTAC linker for targeted protein degradation
  • Click chemistry reagent for efficient conjugation
Catalog No. 50-003-57122 Supplier Medchemexpress LLC Supplier No. 25MGHY140911
Biotin-PEG6-azide is a biotin-labeled, PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and functions as a click chemistry reagent, facilitating targeted protein degradation applications.
  • Biotin-labeled for detection and purification
  • PEG-based for improved solubility and reduced immunogenicity
  • PROTAC linker for targeted protein degradation
  • Click chemistry reagent for efficient conjugation