Learn More
Medchemexpress LLC Biotin-PEG6-azide | 1085938-09-5 | 99.9% | C24H44N6O8S | 50 MG
SDP
Catalog No. 5000353139
Encompass_Preferred
Biotin-PEG6-azide is a biotin-labeled, PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53139 Supplier Medchemexpress LLC Supplier No. HY14091150MG
Biotin-PEG6-azide is a biotin-labeled, PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions