Learn More
Medchemexpress LLC Biotin-PEG6-Mal | 1808990-66-0 | 99.0% | C31H51N5O11S | 100 MG
SDP
Catalog No. 5000354565
Encompass_Preferred
Biotin-PEG6-Mal is a biotin-labeled, PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs function by connecting two different ligands: one for an E3 ubiquitin ligase and another for the target protein. This product is for research use only.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-54565 Supplier Medchemexpress LLC Supplier No. HY140908100MG
Biotin-PEG6-Mal is a biotin-labeled, PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs function by connecting two different ligands: one for an E3 ubiquitin ligase and another for the target protein. This product is for research use only.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only