Learn More
Medchemexpress LLC Biotin-PEG6-Mal | 1808990-66-0 | 99.0% | C31H51N5O11S | 50 MG
SDP
Catalog No. 5000355000
Encompass_Preferred
Biotin-PEG6-Mal is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs selectively degrade target proteins by utilizing the ubiquitin-proteasome system
Catalog No. 50-003-55000 Supplier Medchemexpress LLC Supplier No. HY14090850MG
Biotin-PEG6-Mal is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs selectively degrade target proteins by utilizing the ubiquitin-proteasome system