Learn More
Medchemexpress LLC Biotin-PEG6-Mal | 1808990-66-0 | MFCD28015769 | 99.0% | C31H51N5O11S | 25 MG
SDP
Catalog No. 5000352662
Encompass_Preferred
Biotin-PEG6-Mal is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a target protein ligand.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52662 Supplier Medchemexpress LLC Supplier No. 25MGHY140908
Biotin-PEG6-Mal is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a target protein ligand.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins