Learn More
Medchemexpress LLC Biotin-PEG6-NHS ester | 2055045-04-8 | 98.0% | C29H48N4O12S | 25 MG
SDP
Catalog No. 5000352636
Encompass_Preferred
Biotin-PEG6-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
  • Connects ligands for E3 ubiquitin ligase and target proteins
Catalog No. 50-003-52636 Supplier Medchemexpress LLC Supplier No. 25MGHY140890
Biotin-PEG6-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
  • Connects ligands for E3 ubiquitin ligase and target proteins