Learn More
Medchemexpress LLC Biotin-PEG6-NHS ester | 2055045-04-8 | 98.0% | C29H48N4O12S | 50 MG
SDP
Catalog No. 5000355071
Encompass_Preferred
Biotin-PEG6-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. They function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs target E3 ubiquitin ligase and a protein of interest
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Intended for research use only
Catalog No. 50-003-55071 Supplier Medchemexpress LLC Supplier No. HY14089050MG
Biotin-PEG6-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. They function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs target E3 ubiquitin ligase and a protein of interest
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Intended for research use only