Learn More
Medchemexpress LLC Biotin-PEG6-silane | 00-00-0 | MFCD | 98.0% | C34H66N4O12SSi | 10MG
SDP
Catalog No. 5000260547
Encompass_Preferred
Biotin-PEG6-Silane is a biotinylated polyethylene glycol (PEG6) silane linker supplied as a solid for research use. It offers a flexible spacer with an affinity tag and a silane reactive group for incorporation into conjugates or for surface functionalization in chemical biology and PROTAC workflows.
  • Provides a PEG6 spacer between functional groups.
  • Contains a biotin affinity tag for streptavidin binding.
  • Includes a silane group for covalent attachment to hydroxylated surfaces.
  • High purity (98.0%) for reliable synthetic performance.
  • Supplied as a solid with defined cold storage recommendations.
Catalog No. 50-002-60547 Supplier Medchemexpress LLC Supplier No. HY13841910MG
Biotin-PEG6-Silane is a biotinylated polyethylene glycol (PEG6) silane linker supplied as a solid for research use. It offers a flexible spacer with an affinity tag and a silane reactive group for incorporation into conjugates or for surface functionalization in chemical biology and PROTAC workflows.
  • Provides a PEG6 spacer between functional groups.
  • Contains a biotin affinity tag for streptavidin binding.
  • Includes a silane group for covalent attachment to hydroxylated surfaces.
  • High purity (98.0%) for reliable synthetic performance.
  • Supplied as a solid with defined cold storage recommendations.