Learn More
Medchemexpress LLC Biotin-PEG6-Thalidomide | 2144775-48-2 | 98.3% | 791.91 | 5 MG
SDP
Catalog No. 5000364354
Encompass_Preferred
Biotin-PEG6-Thalidomide is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-64354 Supplier Medchemexpress LLC Supplier No. 5MGHY140942
Biotin-PEG6-Thalidomide is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only