Learn More
Medchemexpress LLC Biotin-PEG6-Thalidomide | 2144775-48-2 | 98.3% | C37H53N5O12S | 25 MG
SDP
Catalog No. 5000352527
Encompass_Preferred
Biotin-PEG6-Thalidomide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs contain two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. They leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets E3 ligase ligand-linker conjugates
  • Targets Cereblon
  • For research use only
Catalog No. 50-003-52527 Supplier Medchemexpress LLC Supplier No. 25MGHY140942
Biotin-PEG6-Thalidomide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs contain two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. They leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets E3 ligase ligand-linker conjugates
  • Targets Cereblon
  • For research use only