Learn More
Medchemexpress LLC Biotin-PEG6-thalidomide | 2144775-48-2 | MFCD31561114 | 98.3% | C37H53N5O12S | 10MG
SDP
Catalog No. 5000261263
Encompass_Preferred
Biotin-PEG6-thalidomide is a biotinylated polyethylene glycol (PEG6)-linked thalidomide derivative used as a PROTAC linker and cereblon (CRBN) affinity probe for targeted protein degradation research. It provides a hydrophilic spacer and a biotin handle to facilitate synthesis, biochemical assays, and affinity-based capture of protein complexes. Supplied for research use only.
  • Provides a PEG6 spacer to increase solubility and flexibility
  • Contains a thalidomide-derived moiety for cereblon binding
  • Includes a biotin tag for affinity capture and pull-down assays
  • Suitable for PROTAC synthesis and CRBN-binding studies
  • Compatible with common organic solvents and assay conditions
Catalog No. 50-002-61263 Supplier Medchemexpress LLC Supplier No. HY14094210MG
Biotin-PEG6-thalidomide is a biotinylated polyethylene glycol (PEG6)-linked thalidomide derivative used as a PROTAC linker and cereblon (CRBN) affinity probe for targeted protein degradation research. It provides a hydrophilic spacer and a biotin handle to facilitate synthesis, biochemical assays, and affinity-based capture of protein complexes. Supplied for research use only.
  • Provides a PEG6 spacer to increase solubility and flexibility
  • Contains a thalidomide-derived moiety for cereblon binding
  • Includes a biotin tag for affinity capture and pull-down assays
  • Suitable for PROTAC synthesis and CRBN-binding studies
  • Compatible with common organic solvents and assay conditions