Learn More
Medchemexpress LLC Biotin-PEG7-amine | 1334172-76-7 | MFCD21363308 | 98.0% | C26H50N4O9S | 10MG
SDP
Catalog No. 5000260316
Encompass_Preferred
Biotin-PEG7-amine is a PEG-based biotinylation reagent containing a seven-unit polyethylene glycol spacer terminating in a primary amine. It is used as a linker in PROTAC synthesis and for bioconjugation to introduce a biotin tag for affinity capture and detection.
  • PEG7 spacer improves solubility and reduces steric hindrance.
  • Terminal primary amine enables coupling to activated carboxyl groups or NHS esters.
  • Biotin tag allows efficient affinity capture using streptavidin or avidin systems.
  • Compatible with standard peptide- and small-molecule coupling chemistries.
  • High purity supports synthetic and analytical applications.
Catalog No. 50-002-60316 Supplier Medchemexpress LLC Supplier No. HY14089810MG
Biotin-PEG7-amine is a PEG-based biotinylation reagent containing a seven-unit polyethylene glycol spacer terminating in a primary amine. It is used as a linker in PROTAC synthesis and for bioconjugation to introduce a biotin tag for affinity capture and detection.
  • PEG7 spacer improves solubility and reduces steric hindrance.
  • Terminal primary amine enables coupling to activated carboxyl groups or NHS esters.
  • Biotin tag allows efficient affinity capture using streptavidin or avidin systems.
  • Compatible with standard peptide- and small-molecule coupling chemistries.
  • High purity supports synthetic and analytical applications.