Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG7-maleimide | 1898221-65-2 | 99.4% | 745.88 g/mol | C33H55N5O12S | 5 MG
SDP

Catalog No. 5000204497
Encompass_Preferred

Biotin-PEG7-Maleimide is a biotinylation reagent that reacts specifically with thiol (-SH) groups via a maleimide linker. It is used to attach biotin to thiol-containing molecules and as a linker in drug-linker conjugates such as antibody-drug conjugates (ADCs).

  • Reacts selectively with thiol groups (-SH).
  • Contains a PEG7 spacer to improve solubility and flexibility.
  • Facilitates biotinylation for detection, pull-down, and affinity capture.
  • Suitable for drug-linker conjugates, including ADC development.
  • Molecular weight 745.88 g/mol.
  • CAS number 1898221-65-2.
  • Purity 99.4%.

Catalog No. 50-002-04497 Supplier Medchemexpress LLC Supplier No. HY1563775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG7-Maleimide is a biotinylation reagent that reacts specifically with thiol (-SH) groups via a maleimide linker. It is used to attach biotin to thiol-containing molecules and as a linker in drug-linker conjugates such as antibody-drug conjugates (ADCs).

  • Reacts selectively with thiol groups (-SH).
  • Contains a PEG7 spacer to improve solubility and flexibility.
  • Facilitates biotinylation for detection, pull-down, and affinity capture.
  • Suitable for drug-linker conjugates, including ADC development.
  • Molecular weight 745.88 g/mol.
  • CAS number 1898221-65-2.
  • Purity 99.4%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.