Learn More
Medchemexpress LLC Biotin-PEG8-acid | 2143964-62-7 | 99.8% | C29H53N3O12S | 50 MG
SDP
Catalog No. 5000356929
Encompass_Preferred
Biotin-PEG8-acid is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • Appears as a white to off-white solid
  • Molecular weight: 667.81
Catalog No. 50-003-56929 Supplier Medchemexpress LLC Supplier No. HY14049250MG
Biotin-PEG8-acid is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • Appears as a white to off-white solid
  • Molecular weight: 667.81