Learn More
Medchemexpress LLC Biotin-PEG8-alcohol | 2182601-20-1 | 97.16% | C26H49N3O10S | 50 MG
SDP
Catalog No. 5000355165
Encompass_Preferred
Biotin-PEG8-alcohol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55165 Supplier Medchemexpress LLC Supplier No. HY14089450MG
Biotin-PEG8-alcohol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins