Learn More
Medchemexpress LLC Biotin-PEG8-alcohol | 2182601-20-1 | 97.2% | C26H49N3O10S | 100 MG
SDP
Catalog No. 5000355111
Encompass_Preferred
Biotin-PEG8-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system, enabling the selective degradation of target proteins. This compound serves as a crucial component for developing targeted therapy drugs.
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Crucial component for developing targeted therapy drugs
Catalog No. 50-003-55111 Supplier Medchemexpress LLC Supplier No. HY140894100MG
Biotin-PEG8-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system, enabling the selective degradation of target proteins. This compound serves as a crucial component for developing targeted therapy drugs.
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Crucial component for developing targeted therapy drugs