Learn More
Medchemexpress LLC Biotin-PEG8-amine | 2183447-27-8 | 99.8% | C28H54N4O10S | 250 MG
SDP
Catalog No. 5000352881
Encompass_Preferred
Biotin-PEG8-amine is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled, PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • For research use only.
  • Appearance: solid, white to off-white.
Catalog No. 50-003-52881 Supplier Medchemexpress LLC Supplier No. 250MGHY140899
Biotin-PEG8-amine is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled, PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • For research use only.
  • Appearance: solid, white to off-white.