Learn More
Medchemexpress LLC Biotin-PEG8-azide | 2363756-34-5 | >98.0% | C32H59N7O11S | 25 MG
SDP
Catalog No. 5000283178
Encompass_Preferred
Biotin-PEG8-azide is a biotinylated polyethylene glycol (PEG) linker bearing an azide functional group for click chemistry. The PEG8 spacer improves solubility and reduces steric hindrance; the azide enables covalent coupling to alkyne-containing molecules via azide-alkyne cycloaddition, and the biotin moiety allows high-affinity capture and detection with streptavidin/avidin systems. Intended for research use in bioconjugation and surface modification applications.
  • Enables Cu(I)-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • Provides biotin tag for streptavidin/avidin capture and detection.
  • PEG8 spacer increases solubility and reduces steric interference.
  • High purity suitable for research bioconjugation workflows.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-002-83178 Supplier Medchemexpress LLC Supplier No. HYW19082425MG
Biotin-PEG8-azide is a biotinylated polyethylene glycol (PEG) linker bearing an azide functional group for click chemistry. The PEG8 spacer improves solubility and reduces steric hindrance; the azide enables covalent coupling to alkyne-containing molecules via azide-alkyne cycloaddition, and the biotin moiety allows high-affinity capture and detection with streptavidin/avidin systems. Intended for research use in bioconjugation and surface modification applications.
  • Enables Cu(I)-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • Provides biotin tag for streptavidin/avidin capture and detection.
  • PEG8 spacer increases solubility and reduces steric interference.
  • High purity suitable for research bioconjugation workflows.
  • Stable as a powder at -20°C for long-term storage.