Learn More
Medchemexpress LLC Biotin-PEG8-NHS ester | 2143968-03-8 | MFCD27977502 | 99.9% | C33H56N4O14S | 10MG
SDP
Catalog No. 5000260467
Encompass_Preferred
Biotin-PEG8-NHS ester is a PEG-based heterobifunctional linker that combines a biotin affinity tag with an N-hydroxysuccinimide (NHS) ester to enable efficient conjugation to primary amines. It is suitable for biotinylation of peptides, proteins, and small molecules, and for use as a linker in targeted protein degradation research and affinity capture workflows.
  • Enables efficient conjugation to primary amines via NHS ester chemistry.
  • Provides a biotin tag for streptavidin-based capture and detection.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity for reproducible conjugation and downstream analysis.
  • Suitable for synthesis of linkers and bioconjugates in research applications.
Catalog No. 50-002-60467 Supplier Medchemexpress LLC Supplier No. HY14089110MG
Biotin-PEG8-NHS ester is a PEG-based heterobifunctional linker that combines a biotin affinity tag with an N-hydroxysuccinimide (NHS) ester to enable efficient conjugation to primary amines. It is suitable for biotinylation of peptides, proteins, and small molecules, and for use as a linker in targeted protein degradation research and affinity capture workflows.
  • Enables efficient conjugation to primary amines via NHS ester chemistry.
  • Provides a biotin tag for streptavidin-based capture and detection.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity for reproducible conjugation and downstream analysis.
  • Suitable for synthesis of linkers and bioconjugates in research applications.