Learn More
Medchemexpress LLC Biotin pentafluorophenyl ester | 120550-35-8 | 97.0% | 100 MG
SDP
Catalog No. 5000233070
Encompass_Preferred
Biotin pentafluorophenyl ester is a biotinylated activated ester used to introduce a biotin affinity tag through reaction with nucleophiles. It serves as an alkyl/ether-based PROTAC linker and reagent for biotinylation in chemical biology and proteomics workflows.
  • Activated pentafluorophenyl (PFP) ester for efficient coupling to amines and alcohols.
  • Introduces a biotin handle for streptavidin-based capture and detection.
  • Useful in PROTAC linker assembly, probe synthesis, and affinity enrichment.
  • Supplied as a white to off-white solid with molecular weight 410.36 g·mol⁻¹.
Catalog No. 50-002-33070 Supplier Medchemexpress LLC Supplier No. HY119343100MG
Biotin pentafluorophenyl ester is a biotinylated activated ester used to introduce a biotin affinity tag through reaction with nucleophiles. It serves as an alkyl/ether-based PROTAC linker and reagent for biotinylation in chemical biology and proteomics workflows.
  • Activated pentafluorophenyl (PFP) ester for efficient coupling to amines and alcohols.
  • Introduces a biotin handle for streptavidin-based capture and detection.
  • Useful in PROTAC linker assembly, probe synthesis, and affinity enrichment.
  • Supplied as a white to off-white solid with molecular weight 410.36 g·mol⁻¹.