Learn More
AdipoGen (+)-Biotin-PFP-ester (100 mg)
SDP
Supplier:  AdipoGen CDXB0308M100
Encompass_Preferred
Chemical. CAS: 120550-35-8. Formula: C16H15F5N2O3S. MW: 410.36. Amine-reactive biotinylation agent.
Catalog No. 50-205-4311