Learn More
Medchemexpress LLC Biotin-PFP ester | 120550-35-8 | 97.0% | 1 G
SDP
Catalog No. 5000234282
Encompass_Preferred
Biotin-PFP ester is a biotinylated pentafluorophenyl (PFP) ester reagent used as an alkyl/ether-based PROTAC linker for the synthesis of PROTAC molecules. It provides an activated ester for coupling to amine-containing ligands and a biotin affinity tag for purification or detection.
  • Provides a biotin handle for affinity capture and detection.
  • Features a PFP-activated ester for efficient coupling to amine-containing ligands.
  • Suitable as an alkyl/ether-based linker in PROTAC synthesis workflows.
  • Offered in multiple laboratory-scale pack sizes for convenient use.
Catalog No. 50-002-34282 Supplier Medchemexpress LLC Supplier No. HY1193431G
Biotin-PFP ester is a biotinylated pentafluorophenyl (PFP) ester reagent used as an alkyl/ether-based PROTAC linker for the synthesis of PROTAC molecules. It provides an activated ester for coupling to amine-containing ligands and a biotin affinity tag for purification or detection.
  • Provides a biotin handle for affinity capture and detection.
  • Features a PFP-activated ester for efficient coupling to amine-containing ligands.
  • Suitable as an alkyl/ether-based linker in PROTAC synthesis workflows.
  • Offered in multiple laboratory-scale pack sizes for convenient use.