Learn More
Medchemexpress LLC Biotin-PFP ester | 120550-35-8 | 97.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC HY119343250MG
Encompass_Preferred
Biotin-PFP ester is a pentafluorophenyl-activated biotin reagent for bioconjugation and linker chemistry. The activated ester reacts efficiently with primary amines to form stable amide bonds, enabling biotin tagging for affinity capture, detection, and proteomics workflows. Supplied as a research-grade solid suitable for small-scale synthesis.
  • Activated pentafluorophenyl ester for efficient amide coupling
  • Provides a biotin tag for affinity purification and detection
  • Compatible with conjugation to primary amines under mild conditions
  • High purity for reproducible synthetic and proteomics results
  • Packaged for research-scale use and storage
Catalog No. 50-002-34713