Learn More
Echelon Biosciences Research Labs BIOTIN-PHOSPHATIDYLSERINE 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs L31B1650UG
Encompass
Biotin-Phosphatidylserine is a biotinylated conjugate of phosphatidylserine (PS), a negatively charged phospholipid located in the inner leaflet of the cell membrane. During apoptosis PS is no longer restricted to the inner leaflet of the plasma membrane and migrates to the outer leaflet. Recognition of extra-leaflet PS by Anexin V is the basis of popular apoptosis assays. Similarly, during platelet activation, PS is translocated to the outer side where it facilitates assembly of the prothrombinase complex. This item is non returnable
Catalog No. NC0206340