Learn More
AVIDITY LLC BIOTIN PROT.LIGASE 40UG 2X20
Catalog No. NC9918597
Encompass
Biotin Protein Ligase 2 vials of 20 microgram (40 microgram);Concentration: 1 mg/ml. Activity: 5000 Units/microgram
Catalog No. NC9918597 Supplier AVIDITY LLC Supplier No. BIRA500
Biotin Protein Ligase 2 vials of 20 microgram (40 microgram);Concentration: 1 mg/ml. Activity: 5000 Units/microgram