Learn More
Genscript Corporation Antibody Protein Protein Analysis Protein Analysis 2 MG 1/EA
Supplier:  Genscript Corporation M00095
Encompass
Staphylococcal Protein A binds strongly and specifically to the Fc region of immunoglobulin molecules particularly IgG. The product contains five high-affinity binding sites capable of interacting with the Fc region of IgG in humans rabbits pigs and other species. Biotin-Protein A which is produced by covalently linking biotin to protein A can be used in immunodetections such as immunoblotting (western or dot blot) ELISA and immunohistochemistry.
Catalog No. 50-252-9965