Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin Protein Labeling Ki
Catalog No. 502905489
Encompass
for labeling of proteins with biotin.free amino acids of the protein to be labeled react with D-biotinoyl-epsilon-aminocaproic acid-N-hydroxy-succinimide ester (biotin-7-NHS) by forming a stable amino bond. Non reacted biotin-7-NHS is separated on a Sephadex G-25 column.
Catalog No. 50-290-5489 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11418165001
for labeling of proteins with biotin.free amino acids of the protein to be labeled react with D-biotinoyl-epsilon-aminocaproic acid-N-hydroxy-succinimide ester (biotin-7-NHS) by forming a stable amino bond. Non reacted biotin-7-NHS is separated on a Sephadex G-25 column.