Learn More
Echelon Biosciences Research Labs Biotin-PtdIns 10 ug
SDP
Catalog No. 50143985
Encompass
Biotin Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with biotin at the sn-1 position. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-143-985 Supplier Echelon Biosciences Research Labs Supplier No. C00B610UG
Biotin Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with biotin at the sn-1 position. This item is non returnable