Learn More
Echelon Biosciences Research Labs Biotin-PtdIns 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs C00B650UG
Encompass
Biotin Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with biotin at the sn-1 position. This item is non returnable
Catalog No. 50-143-986