Learn More
Apexbio Technology LLC Biotin Rabbit Anti-Goat IgG (H+L) Antibody 2X1ml
SDP
Catalog No. 5000096952
Encompass_Preferred
Biotin Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal secondary antibody targeting goat IgG heavy and light chains It is designed to specifically bind to goat IgG via antigen-antibody interactions thereby enabling detection and quantification of goat-derived antibodies Biotin Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity primarily through selective binding to goat IgG (H L) chains and is conjugated to biotin for subsequent signal amplification via streptavidin/avidin systems Based on these properties Biotin Rabbit Anti-Goat IgG HplusL Antibody holds research potential in ELISA Western blot immunohistochemistry and immunofluorescence applications
Catalog No. 50-000-96952 Supplier Apexbio Technology LLC Supplier No. K12202
Biotin Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal secondary antibody targeting goat IgG heavy and light chains It is designed to specifically bind to goat IgG via antigen-antibody interactions thereby enabling detection and quantification of goat-derived antibodies Biotin Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity primarily through selective binding to goat IgG (H L) chains and is conjugated to biotin for subsequent signal amplification via streptavidin/avidin systems Based on these properties Biotin Rabbit Anti-Goat IgG HplusL Antibody holds research potential in ELISA Western blot immunohistochemistry and immunofluorescence applications