Learn More
Seracare Life Sciences Inc BIOTIN-RABBIT ANTI-VIBRIO
Supplier:  Seracare Life Sciences Inc 53600027
Encompass_Preferred
Protein G purified BacTrace antibody to Vibrio made in rabbit labeled with biotin. Antibody is broadly reactive for Vibrio.
Catalog No. 50-672-90