Learn More
Immunology Consultants anti-Goat IgG h+l Antibody (Rabbit) - Biotin Conjugated
SDP
Catalog No. 502248873
Encompass_Preferred
RGHL‑50B is a biotin‑conjugated rabbit antibody specific for goat IgG heavy and light chains, enabling streptavidin‑based detection with enhanced sensitivity. This reagent is ideal for ELISA, Western blotting, and amplified detection workflows requiring strong, selective binding to goat immunoglobulins. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50B offers versatile, high‑performance detection capabilities.
This item is not returnable. View return policy
Catalog No. 50-224-8873 Supplier Immunology Consultants Supplier No. RGHL50B
RGHL‑50B is a biotin‑conjugated rabbit antibody specific for goat IgG heavy and light chains, enabling streptavidin‑based detection with enhanced sensitivity. This reagent is ideal for ELISA, Western blotting, and amplified detection workflows requiring strong, selective binding to goat immunoglobulins. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50B offers versatile, high‑performance detection capabilities.