Learn More
Seracare Life Sciences Inc BIOTIN-SA HRP KIT RB PRI AB
Catalog No. 5067566
Encompass_Preferred
HistoMark Biotin Streptavidin-HRP Systems provide rapid, precise localization of cell surface and intracellular antigens in frozen or paraffin-embedded tissue, cytospins and touch preparations. These systems also facilitate double or triple labeleing experiments, as detection can be performed simultaneously with primary antibodies of different animal species. Streptavidin demonstrates less non-specific binding than avidin. Biotinylated anti-rabbit antibody is human serum adsorbed to minimize cross-reactivity.
This item is not returnable. View return policy
Catalog No. 50-675-66 Supplier Seracare Life Sciences Inc Supplier No. 55200024
HistoMark Biotin Streptavidin-HRP Systems provide rapid, precise localization of cell surface and intracellular antigens in frozen or paraffin-embedded tissue, cytospins and touch preparations. These systems also facilitate double or triple labeleing experiments, as detection can be performed simultaneously with primary antibodies of different animal species. Streptavidin demonstrates less non-specific binding than avidin. Biotinylated anti-rabbit antibody is human serum adsorbed to minimize cross-reactivity.