Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-sar-oh | 154024-76-7 | 315.39 | C13H21N3O4S | 1 G
SDP

Catalog No. 5000203078
Encompass_Preferred

Biotin-sar-oh is a cleavable antibody-drug conjugate (ADC) linker bearing a biotin functionality, used in the synthesis of antibody-drug conjugates and probe development.

  • Cleavable ADC linker for conjugation and release strategies.
  • Biotinylated for affinity capture and detection.
  • Molecular formula: C13H21N3O4S; molecular weight: 315.39.
  • Available pack sizes: 250 mg and 1 g.
  • CAS number: 154024-76-7.

Catalog No. 50-002-03078 Supplier Medchemexpress LLC Supplier No. HYW0961271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-sar-oh is a cleavable antibody-drug conjugate (ADC) linker bearing a biotin functionality, used in the synthesis of antibody-drug conjugates and probe development.

  • Cleavable ADC linker for conjugation and release strategies.
  • Biotinylated for affinity capture and detection.
  • Molecular formula: C13H21N3O4S; molecular weight: 315.39.
  • Available pack sizes: 250 mg and 1 g.
  • CAS number: 154024-76-7.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.